COMH4211B158 Combi-Blocks, Inc.

3-Chloro-2-cyclopentylsulfanyl-pyridine

  • MDL number MFCD31557916
  • Cas Number : 1623886-01-0
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3898

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3898
Availability details for
Quantity
  • Availability
  • Add to Cart