COMH4211B159 Combi-Blocks, Inc.

Ethyl 3-(5-amino-3-tert-butyl-1h-pyrazol-1-yl)benzoate

  • MDL number MFCD28404255
  • Cas Number : 725685-92-7
  • Molecular Weight : 287.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3908
Availability details for
Quantity
  • Availability
  • Add to Cart