COMH4211B15A Combi-Blocks, Inc.

N,N-Diethyl-5-iodo-2-methyl-benzamide

  • MDL number MFCD31381589
  • Cas Number : 2203717-19-3
  • Molecular Weight : 317.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3909

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3909
Availability details for
Quantity
  • Availability
  • Add to Cart