COMH4211B15B Combi-Blocks, Inc.

5-Methyl-2-(2,2,2-trifluoro-ethoxy)-benzoic acid

  • MDL number MFCD08696486
  • Cas Number : 937602-67-0
  • Molecular Weight : 234.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3919
Availability details for
Quantity
  • Availability
  • Add to Cart