COMH4211B15E Combi-Blocks, Inc.

5-Bromo-2-ethoxy-4-methyl-benzoic acid ethyl ester

  • MDL number MFCD25459853
  • Cas Number : 474555-42-5
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3924

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3924
Availability details for
Quantity
  • Availability
  • Add to Cart