COMH4211B15F Combi-Blocks, Inc.

(R)-N-(2-(4-Isopropyl-4,5-dihydrooxazol-2-yl)-6-methylphenyl)methanesulfonamide

  • MDL number MFCD31381702
  • Cas Number : 871360-40-6
  • Molecular Weight : 296.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3925

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3925
Availability details for
Quantity
  • Availability
  • Add to Cart