COMH4211B161 Combi-Blocks, Inc.

Trans 4-(4-tert-butoxycarbonylamino-cyclohexyl)-butyric acid

  • MDL number MFCD31579413
  • Cas Number : 2270910-76-2
  • Molecular Weight : 285.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3927
Availability details for
Quantity
  • Availability
  • Add to Cart