COMH4211B163 Combi-Blocks, Inc.

4-Bromo-3-fluoro-n-(2-hydroxyethyl)benzamide

  • MDL number MFCD16084811
  • Cas Number : 1248358-31-7
  • Molecular Weight : 262.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3932
Availability details for
Quantity
  • Availability
  • Add to Cart