COMH4211B164 Combi-Blocks, Inc.

2-(Cyclopentylsulfanyl)pyrimidine-5-carboxylic acid

  • MDL number MFCD31583360
  • Cas Number : 1879597-19-9
  • Molecular Weight : 224.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3933
Availability details for
Quantity
  • Availability
  • Add to Cart