COMH4211B165 Combi-Blocks, Inc.

1-(3-Fluoro-4-propoxy-phenyl)-ethylamine

  • MDL number MFCD11619318
  • Cas Number : 1094760-74-3
  • Molecular Weight : 197.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3937
Availability details for
Quantity
  • Availability
  • Add to Cart