COMH4211B167 Combi-Blocks, Inc.

1-[3-Chloro-4-(2-methoxy-ethoxy)-phenyl]-ethylamine

  • MDL number MFCD16315256
  • Cas Number : 1226114-35-7
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3942

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3942
Availability details for
Quantity
  • Availability
  • Add to Cart