COMH4211B168 Combi-Blocks, Inc.

6-Chloro-n-(cyclopropylmethyl)pyrimidin-4-amine

  • MDL number MFCD14611189
  • Cas Number : 928650-17-3
  • Molecular Weight : 183.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3944

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3944
Availability details for
Quantity
  • Availability
  • Add to Cart