COMH4211B169 Combi-Blocks, Inc.

1-(Cyclopropylmethoxy)-4-iodobenzene

  • MDL number MFCD14659233
  • Cas Number : 411229-57-7
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(cyclopropylmethoxy)-4-iodobenzene
Inchi Code 1S/C10H11IO/c11-9-3-5-10(6-4-9)12-7-8-1-2-8/h3-6,8H,1-2,7H2
Inchi Key DWCTVGYGXSIFST-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3945

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3945
Availability details for
Quantity
  • Availability
  • Add to Cart