COMH4211B16B Combi-Blocks, Inc.

1-(Cyclopropanesulfonyl)-4-methylpiperidin-4-amine

  • MDL number MFCD30613627
  • Cas Number : 1866740-92-2
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3948
Availability details for
Quantity
  • Availability
  • Add to Cart