COMH4211B16C Combi-Blocks, Inc.

2-(4-Bromophenyl)-n-(cyclopropylmethyl)ethanamine

  • MDL number MFCD12816034
  • Cas Number : 1178914-20-9
  • Molecular Weight : 254.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-bromophenyl)-N-(cyclopropylmethyl)ethanamine
Inchi Code 1S/C12H16BrN/c13-12-5-3-10(4-6-12)7-8-14-9-11-1-2-11/h3-6,11,14H,1-2,7-9H2
Inchi Key LHMTZWVKFUHTSZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3950
Availability details for
Quantity
  • Availability
  • Add to Cart