COMH4211B16E Combi-Blocks, Inc.

Dihydro donepezil

  • MDL number MFCD18382291
  • Cas Number : 120012-04-6
  • Molecular Weight : 381.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3954

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3954
Availability details for
Quantity
  • Availability
  • Add to Cart