COMH4211B16F Combi-Blocks, Inc.

(3-Methoxy-4-nitrophenyl)hydrazine

  • MDL number MFCD18861001
  • Cas Number : 648917-64-0
  • Molecular Weight : 183.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-methoxy-4-nitrophenyl)hydrazine
Inchi Code 1S/C7H9N3O3/c1-13-7-4-5(9-8)2-3-6(7)10(11)12/h2-4,9H,8H2,1H3
Inchi Key PCZJMSVFHJMNSC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3956

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3956
Availability details for
Quantity
  • Availability
  • Add to Cart