COMH4211B170 Combi-Blocks, Inc.

2-Chloro-3-(trifluoromethyl)pyridine 1-oxide

  • MDL number MFCD14581996
  • Cas Number : 164464-57-7
  • Molecular Weight : 197.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3959

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3959
Availability details for
Quantity
  • Availability
  • Add to Cart