COMH4211B171 Combi-Blocks, Inc.

1-(4-Bromo-2-isobutoxy-phenyl)-ethanone

  • MDL number MFCD24110647
  • Cas Number : 1556600-65-7
  • Molecular Weight : 271.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3963

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3963
Availability details for
Quantity
  • Availability
  • Add to Cart