COMH4211B172 Combi-Blocks, Inc.

(5-Bromo-pyrazin-2-yl)-(3-cyclopentyloxy-4-methoxy-benzyl)-amine

  • MDL number MFCD31579497
  • Cas Number : 945978-39-2
  • Molecular Weight : 378.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3966

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3966
Availability details for
Quantity
  • Availability
  • Add to Cart