COMH4211B173 Combi-Blocks, Inc.

2,6-Dichloro-n-(cyclopropylmethyl)aniline

  • MDL number MFCD21242434
  • Cas Number : 87425-12-5
  • Molecular Weight : 216.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3971
Availability details for
Quantity
  • Availability
  • Add to Cart