COMH4211B175 Combi-Blocks, Inc.

6-(4-Methoxy-benzyloxy)-3,4-dihydro-2h-naphthalen-1-one

  • MDL number MFCD31583117
  • Cas Number : 139149-06-7
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3975

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3975
Availability details for
Quantity
  • Availability
  • Add to Cart