COMH4211B176 Combi-Blocks, Inc.

5-Ethynyl-1-(tetrahydro-2h-pyran-2-yl)-1h-indazole

  • MDL number MFCD28142747
  • Cas Number : 1365888-99-8
  • Molecular Weight : 226.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3976
Availability details for
Quantity
  • Availability
  • Add to Cart