COMH4211B177 Combi-Blocks, Inc.

3-(6-Methoxy-naphthalen-2-yl)-2-methyl-propionic acid

  • MDL number MFCD24760783
  • Cas Number : 113591-52-9
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3977

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3977
Availability details for
Quantity
  • Availability
  • Add to Cart