COMH4211B178 Combi-Blocks, Inc.

[3-(Benzyloxy)-4-bromophenyl]methanol

  • MDL number MFCD09038130
  • Cas Number : 540779-36-0
  • Molecular Weight : 293.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3979

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3979
Availability details for
Quantity
  • Availability
  • Add to Cart