COMH4211B179 Combi-Blocks, Inc.

4-Methylphenyl 2,4,5-trifluorobenzoate

  • MDL number MFCD30830210
  • Cas Number : 1354960-66-9
  • Molecular Weight : 266.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3980
Availability details for
Quantity
  • Availability
  • Add to Cart