COMH4211B17A Combi-Blocks, Inc.

4-(4-Bromo-3,5-dimethylphenoxy)-2-methyl-2-butanol

  • MDL number MFCD28130138
  • Cas Number : 1294453-62-5
  • Molecular Weight : 287.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3981

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3981
Availability details for
Quantity
  • Availability
  • Add to Cart