COMH4211B17B Combi-Blocks, Inc.

Chloromethyl heptadecanoate

  • MDL number MFCD31613866
  • Cas Number : 77878-00-3
  • Molecular Weight : 318.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3983

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3983
Availability details for
Quantity
  • Availability
  • Add to Cart