COMH4211B17C Combi-Blocks, Inc.

6-[4-(2-Hydroxy-ethyl)-phenoxy]-nicotinamide

  • MDL number MFCD31618952
  • Cas Number : 676493-96-2
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3985

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3985
Availability details for
Quantity
  • Availability
  • Add to Cart