COMH4211B17D Combi-Blocks, Inc.

4-Bromo-1-(2,2-difluoroethoxy)-2-methylbenzene

  • MDL number MFCD18294105
  • Cas Number : 1339508-59-6
  • Molecular Weight : 251.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3987

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3987
Availability details for
Quantity
  • Availability
  • Add to Cart