COMH4211B17F Combi-Blocks, Inc.

3-(1-Isobutyl-piperidin-4-yl)-propionic acid hydrochloride

  • MDL number MFCD31557822
  • Cas Number : 2203139-74-4
  • Molecular Weight : 249.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3993

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3993
Availability details for
Quantity
  • Availability
  • Add to Cart