COMH4211B182 Combi-Blocks, Inc.

3-Amino-4-nitrobenzeneboronic acid, pinacol ester, n-boc protected

  • MDL number MFCD20036254
  • Cas Number : 1404561-06-3
  • Molecular Weight : 364.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3998

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-3998
Availability details for
Quantity
  • Availability
  • Add to Cart