COMH4211B183 Combi-Blocks, Inc.

4-(4-Amino-2-fluorophenoxy)picolinamide

  • MDL number MFCD12424710
  • Cas Number : 868733-71-5
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4001

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4001
Availability details for
Quantity
  • Availability
  • Add to Cart