COMH4211B186 Combi-Blocks, Inc.

2-Fluoro-4-(2,2,2-trifluoroethoxy)benzaldehyde

  • MDL number MFCD30486346
  • Cas Number : 1588507-91-8
  • Molecular Weight : 222.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4016

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4016
Availability details for
Quantity
  • Availability
  • Add to Cart