COMH4211B188 Combi-Blocks, Inc.

5-Bromo-2-chloro-nicotinic acid 2,5-dioxo-pyrrolidin-1-yl ester

  • MDL number MFCD31539519
  • Cas Number : 2206607-39-6
  • Molecular Weight : 333.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4021

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4021
Availability details for
Quantity
  • Availability
  • Add to Cart