COMH4211B189 Combi-Blocks, Inc.

5-Bromo-2-(oxetan-3-yloxy)benzonitrile

  • MDL number MFCD28663685
  • Cas Number : 1439378-08-1
  • Molecular Weight : 254.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4024
Availability details for
Quantity
  • Availability
  • Add to Cart