COMH4211B1AF Combi-Blocks, Inc.

5-(2,5-Dichlorophenyl)thiophene-2-carboxylic acid

  • MDL number MFCD07375938
  • Cas Number : 1007089-03-3
  • Molecular Weight : 273.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(2,5-dichlorophenyl)-2-thiophenecarboxylic acid
Inchi Code 1S/C11H6Cl2O2S/c12-6-1-2-8(13)7(5-6)9-3-4-10(16-9)11(14)15/h1-5H,(H,14,15)
Inchi Key UOZVUWWGSRUGCR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4025
Availability details for
Quantity
  • Availability
  • Add to Cart