COMH4211B1B0 Combi-Blocks, Inc.

5-Benzyloxy-4-fluoro-2-methyl-benzoic acid

  • MDL number MFCD31579465
  • Cas Number : 2270905-58-1
  • Molecular Weight : 260.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4026

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4026
Availability details for
Quantity
  • Availability
  • Add to Cart