COMH4211B1B1 Combi-Blocks, Inc.

4-(Isopropylamino-methyl)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD21098001
  • Cas Number : 1353989-57-7
  • Molecular Weight : 290.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4027

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4027
Availability details for
Quantity
  • Availability
  • Add to Cart