COMH4211B1B2 Combi-Blocks, Inc.

Methyl 2-bromo-5-iodo-4-methylbenzoate

  • MDL number MFCD30295004
  • Cas Number : 474330-55-7
  • Molecular Weight : 354.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-5-iodo-4-methylbenzoate
Inchi Code 1S/C9H8BrIO2/c1-5-3-7(10)6(4-8(5)11)9(12)13-2/h3-4H,1-2H3
Inchi Key ZALXPDMDZZAHDP-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4030
Availability details for
Quantity
  • Availability
  • Add to Cart