COMH4211B1B3 Combi-Blocks, Inc.

[4-(Piperidin-4-yloxy)-phenyl]-methanol

  • MDL number MFCD14599949
  • Cas Number : 1251336-69-2
  • Molecular Weight : 207.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4031
Availability details for
Quantity
  • Availability
  • Add to Cart