COMH4211B1B7 Combi-Blocks, Inc.

1-[3-Fluoro-4-(2-methoxy-ethoxy)-phenyl]-ethylamine hydrochloride

  • MDL number MFCD31557810
  • Cas Number : 2206608-49-1
  • Molecular Weight : 249.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4046

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4046
Availability details for
Quantity
  • Availability
  • Add to Cart