COMH4211B1B8 Combi-Blocks, Inc.

5-Hydroxy-2-(2-methoxy-ethoxy)-benzamide

  • MDL number MFCD01667477
  • Cas Number : 28173-87-7
  • Molecular Weight : 211.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4048

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4048
Availability details for
Quantity
  • Availability
  • Add to Cart