COMH4211B1B9 Combi-Blocks, Inc.

Methyl 2-(2-cyano-4-fluorophenyl)acetate

  • MDL number MFCD18399059
  • Cas Number : 1261608-35-8
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4049

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4049
Availability details for
Quantity
  • Availability
  • Add to Cart