COMH4211B1BE Combi-Blocks, Inc.

3-(2-Methoxy-phenyl)-2-methyl-propionic acid

  • MDL number MFCD09894706
  • Cas Number : 52427-12-0
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4056

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4056
Availability details for
Quantity
  • Availability
  • Add to Cart