COMH4211B1BF Combi-Blocks, Inc.

1-(3,4-Difluoro-benzyl)-1h-pyrazol-4-ol

  • MDL number MFCD31561512
  • Cas Number : 1603188-42-6
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4057
Availability details for
Quantity
  • Availability
  • Add to Cart