COMH4211B1C0 Combi-Blocks, Inc.

2-(2-Methoxyethoxy)ethyl (3-ethynylphenyl)carbamate

  • MDL number MFCD31579401
  • Cas Number : 1816945-64-8
  • Molecular Weight : 263.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4059

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4059
Availability details for
Quantity
  • Availability
  • Add to Cart