COMH4211B1C2 Combi-Blocks, Inc.

Trans 4-piperidin-1-ylmethyl-cyclohexylamine dihydrochloride

  • MDL number MFCD30377254
  • Cas Number : 2365390-70-9
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4061
Availability details for
Quantity
  • Availability
  • Add to Cart