COMH4211B1C3 Combi-Blocks, Inc.

4-Chloro-2-(4-hydroxy-pyrazol-1-ylmethyl)-benzoic acid methyl ester

  • MDL number MFCD31583339
  • Cas Number : 2270907-90-7
  • Molecular Weight : 266.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4063
Availability details for
Quantity
  • Availability
  • Add to Cart