COMH4211B1C5 Combi-Blocks, Inc.

1-Bromo-2-(2-cyclopropylmethoxy-ethyl)-benzene

  • MDL number MFCD31618991
  • Molecular Weight : 255.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-4067
Availability details for
Quantity
  • Availability
  • Add to Cart